Shimpo FG-3000 Series Digital Force Gauges
Simple, cost-effective tension and compression testing with data logging capability
  • Three modes of operation—track, peak and preset limits—offer maximum flexibility
  • 180° reversible display with tension/compression icons and vertical printed keypad allow for portable or mounting configurations
  • Comparator output function provides easy pass/fail testing system integration capabilities
  • Multilanguage and engineering units selection capability offers additional flexibility
  • Free downloadable data logging/graphing software for quick and easy data analysis
  • Rugged, ergonomic construction withstands demanding applications
These FG-3000 series digital force gauges provide fast sample rates (1000 Hz) and high accuracy for precision measurement results. Units feature three user-selectable modes of operation that include track, peak, and preset limits allowing you to display live readings, record the maximum reading during test, and define high and low set points. Setting the programmable limits provides a quick visual and audible indication if a test passes or fails. In addition, a comparator output enables the simple integration of these gauges into quality systems for repetitive testing such as on production lines.
Large backlit LCD with graphics and icons facilitates quick and easy setup and operation. An analog bar graph displays current readings in comparison to the full-scale range and the pass/fail icons provide an instant response of the testing outcome. The storage symbol indicates when a reading is logged. Use the menu-selectable display orientation function to easily transition from push to pull testing for portable or test stand applications.
The Shimpo FG-3000 series digital force gauges allow you to log up to 500 readings and to view data on the screen, send to the optional printer, or upload via the free downloadable software from Shimp for graphing and statistical analysis capabilities. The wide measuring range (2.2 to 220 lb) is ideal for incoming quality inspection, finished goods testing, R&D, and almost any portable or force stand testing applications.
$350.00 - $404.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Resolution (lbs)
Availability Pricing
Shimpo FG-3003 Digital Force Gauge, 2.2 lbf EW-25301-66 Mfr # FG-3003
COMPARE
2.2 0.001
$350.00USD / Each
Shimpo FG-3005 Digital Force Gauge, 11 lbf EW-25301-67 Mfr # FG-3005
COMPARE
11 0.01
$350.00USD / Each
Shimpo FG-3006 Digital Force Gauge, 22 lbf EW-25301-68 Mfr # FG-3006
COMPARE
22 0.01
$350.00USD / Each
Shimpo FG-3008 Digital Force Gauge, 110 lbf EW-25301-69 Mfr # FG-3008
COMPARE
110 0.1
$404.00USD / Each
Shimpo FG-3009 Digital Force Gauge, 220 lbf EW-25301-70 Mfr # FG-3009
COMPARE
220 0.1
$404.00USD / Each