Shimpo Digital Contact Tachometers
Eliminate inaccurate measurements
  • Rugged die-cast aluminum construction
  • Direct length measurement
  • Extensive speed range (0.10 to 25,000 rpm)
  • Large 5-digit display—choose either LCD or LED
  • Lengthy battery life (40 or 65 hours)
  • Large 10 test memory capacity (selectable to 5 minutes or more)
  • Total revolution display
  • Minimum shaft loading overspeed protection
$270.00 - $450.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Display Type Description
Availability Pricing
Shimpo DT-105A-12KMW Digital Contact Tachometer, LCD Display, 12" Circumference Knurled Aluminum Wheel EW-54500-97 Mfr # DT-105A-12KMW
COMPARE
5-digit LCD Digital Contact Tachometer, LCD Display, 12\" Circumference Knurled Aluminum Wheel
$450.00USD / Each
Shimpo DT-107A Digital Contact Tachometer, LED Display, 6" Standard Wheel EW-98806-29 Mfr # DT-107A
COMPARE
5-digit LED Digital Contact Tachometer, LED Display, 6\" Standard Wheel
$270.00USD / Each
Shimpo DT-107A-S12 Digital Contact Tachometer, LED Display, 12" Standard Wheel EW-98807-37 Mfr # DT-107A-S12
COMPARE
5-digit LED Digital Contact Tachometer, LED Display, 12\" Standard Wheel
$325.00USD / Each