Shimpo Data Logging Digital Force Gauges with Remote Load Cell
Higher capacities and easier positioning via connected load cell
  • Trace, Peak, Auto Peakl, and First Peak selectable modes
  • Program high and low limits for visual/audible indication
  • Store up to 1000 readings in memory for viewing, printing, or downloading
Gauges feature four testing modes: Track for live readings, Peak for displaying the maximum reading, First Peak where only the first peak is displayed, and Auto-Peak where the peak resets after a programmed time. Program high/low set points for go/no go testing; visual and audible alarms indicate pass or fail.
Selectable display shows data numerically or graphically. Log up to 1000 readings for viewing, printing, or download. Multilanguage display allows you to select from English, German, or Chinese. 180 auto-reversible display, tension/compression icons, and dual printed keypad permit use as a portable gauge or mounted upside down on a test stand.
$727.00 - $2,370.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs) Capacity (Newtons) Load Cell
Availability Pricing
Shimpo Digital Force Gauge with S-Beam Load Cell; 220 lb Capacity EW-25303-40 Mfr # FG-7000L-S-1
COMPARE
220 1000 S Beam
$727.00USD / Each
Shimpo Digital Force Gauge with S-Beam Load Cell; 450 lb Capacity EW-25303-41 Mfr # FG-7000L-S-2
COMPARE
450 2000 S Beam
$781.00USD / Each
Shimpo Digital Force Gauge with S-Beam Load Cell; 1100 lb Capacity EW-25303-42 Mfr # FG-7000L-S-5
COMPARE
1100 5000 S Beam
$889.00USD / Each
Shimpo Digital Force Gauge with S-Beam Load Cell; 2250 lb Capacity EW-25303-43 Mfr # FG-7000L-S-10
COMPARE
2250 10000 S Beam
$964.00USD / Each
Shimpo Digital Force Gauge with S-Beam Load Cell; 4500 lb Capacity EW-25303-44 Mfr # FG-7000L-S-20
COMPARE
4500 20000 S Beam
$1,072.00USD / Each
Shimpo Digital Force Gauge with Ring-Type Load Cell; 2250 lb Capacity EW-25303-45 Mfr # FG-7000L-R-10
COMPARE
2250 10000 Ring Type
$1,400.00USD / Each
Shimpo Digital Force Gauge with Ring-Type Load Cell; 4500 lb Capacity EW-25303-46 Mfr # FG-7000L-R-20
COMPARE
4500 20000 Ring Type
$1,481.00USD / Each
Shimpo Digital Force Gauge with Ring-Type Load Cell; 11,000 lb Capacity EW-25303-47 Mfr # FG-7000L-R-50
COMPARE
11000 50000 Ring Type
$1,589.00USD / Each
Shimpo Digital Force Gauge with Ring-Type Load Cell; 22,000 lb Capacity EW-25303-48 Mfr # FG-7000L-R-100
COMPARE
22000 100000 Ring Type
$2,047.00USD / Each
Shimpo Digital Force Gauge with Ring-Type Load Cell; 44,000 lb Capacity EW-25303-49 Mfr # FG-7000L-R-200
COMPARE
44000 200000 Ring Type
$2,370.00USD / Each
Showing 1- 10 of 10