Trajan SGE forte™ Capillary Column for Gas Chromatography, 0.32 mm ID
Offer enhanced endurance without loss of sensitivity
  • SilGuard™ integrated guard column technology provides additional protection by reducing the amount of nonvolatiles entering the analytical column Provides the best possible combination of performance, robustness, reproducibility, low bleed, and inertness Available in a wide variety of phases, IDs, lengths, and film thicknesses
The SGE forte line of GC capillary columns provides the best possible combination of performance, robustness, reproducibility, low bleed, and inertness. It offers a comprehensive range of columns for almost any chromatographic application, including chemical, environmental, food, and fuels.
$445.00 - $1,630.00USD / Each
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Film Thickness (µm) Phase
Availability Pricing
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 0.25 µm; 25 m length EW-17940-39 Mfr # 054059
COMPARE
0.32 0.25 BP1
$868.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 0.25 µm; 15 m length EW-17940-40 Mfr # 054061
COMPARE
0.32 0.25 BP1
$500.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 0.25 µm; 30 m length EW-17940-41 Mfr # 054062
COMPARE
0.32 0.25 BP1
$813.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 0.5 µm; 25 m length EW-17940-42 Mfr # 054065
COMPARE
0.32 0.5 BP1
$901.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 0.5 µm; 30 m length EW-17940-43 Mfr # 054068
COMPARE
0.32 0.5 BP1
$838.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 0.5 µm; 60 m length EW-17940-44 Mfr # 054069
COMPARE
0.32 0.5 BP1
$1,520.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 1 µm; 12 m length EW-17940-45 Mfr # 054070
COMPARE
0.32 1 BP1
$542.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 1 µm; 25 m length EW-17940-46 Mfr # 054071
COMPARE
0.32 1 BP1
$923.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 1 µm; 50 m length EW-17940-47 Mfr # 054072
COMPARE
0.32 1 BP1
$1,520.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 3 µm; 30 m length EW-17940-48 Mfr # 054073
COMPARE
0.32 3 BP1
$813.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 5 µm; 50 m length EW-17940-49 Mfr # 054082
COMPARE
0.32 5 BP1
$1,575.00USD / Each
Trajan SGE GC Column, BP1, 100% Dimethyl Polysiloxane, 0.32 mm x 5 µm; 60 m length EW-17940-50 Mfr # 054085
COMPARE
0.32 5 BP1
$1,465.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.32 mm x 0.25 µm; 12 m length EW-17940-67 Mfr # 054118
COMPARE
0.32 0.25 BPX5
$597.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.32 mm x 0.25 µm; 25 m length EW-17940-68 Mfr # 054119
COMPARE
0.32 0.25 BPX5
$923.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.32 mm x 0.5 µm; 30 m length EW-17940-69 Mfr # 0541205
COMPARE
0.32 0.5 BPX5
$868.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.32 mm x 0.5 µm; 25 m length EW-17940-73 Mfr # 054125
COMPARE
0.32 0.5 BPX5
$976.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.32 mm x 0.5 µm; 50 m length EW-17940-74 Mfr # 054126
COMPARE
0.32 0.5 BPX5
$1,630.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.32 mm x 1 µm; 6 m length EW-17940-75 Mfr # 0541261
COMPARE
0.32 1 BPX5
$445.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.32 mm x 1 µm; 12 m length EW-17940-76 Mfr # 054127
COMPARE
0.32 1 BPX5
$597.00USD / Each
Trajan SGE GC Column, BPX5, 5% Phenyl Polysilphenylene-siloxane, 0.32 mm x 1 µm; 50 m length EW-17940-77 Mfr # 054129
COMPARE
0.32 1 BPX5
$1,575.00USD / Each
Showing 1 - 20 of 50