Scientific Industries Genie Temp-Shaker Orbital Shaking Incubators
Unobstructed 360° view with clear lightweight cover
  • Minimal loss of temperature when door is open due to unique temperature flow
  • Quickly heat chamber to 98.6°F (37°C) in less than 3 minutes
  • Microprocessor-controlled shaking speed and temperature ensure accuracy
These shaking incubators ensure consistent and uniform temperatures of test samples due to unique airflow design. Chamber air is gently and continuously circulated and can easily be controlled via the large, easy-to-read digital display. All models feature variable-speed and temperature multi-vessel orbital shaking. Access samples in three easy ways — door, kickstand (back, left, or right), or removable cover.
The Genie Temp-Shaker 100 orbital shaking incubators feature a 20 to 100 rpm speed range with thermoplastic anti-slip rubber mat. The platform has raised edges to contain any minor spills that may occur during operation. The platform can accommodate up to 2-liter vessels with a 5-pound (2.3 kg) load capacity. Easily increase capacity with stacking tray accessory 51820-21, sold separately.
Achieve higher speeds with the Genie Temp-Shaker 300, which features a 35 to 300 rpm range. Choose from three options. GTS300 with universal ratcheting clamps provides flexibility for using a variety of different size and shape vessels or large vessels on a single platform. The GTS 300 with flask clamps shaker is ideal for accepting various size flasks or beakers (clamps sold separately). Utilize the GTS 300 with adhering mat for use with open containers and vessels that cannot be held by conventional clamps. The patented material provides natural adhering and can be easily cleaned to renew adhering properties.
$3,162.00 - $3,690.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC) Description
Availability Pricing
Scientific Industries Genie Temp-Shaker 100 Orbital Shaking Incubator, 120 VAC; US Standard Plug EW-51603-16 Mfr # SI-G100
COMPARE
100 120 Orbital Shaking Incubator, 120 VAC; US Standard Plug
$3,162.00USD / Each
Scientific Industries Genie Temp-Shaker 300 Orbital Shaking Incubator with Ratcheting Clamps, 120 VAC; US Standard Plug EW-51603-21 Mfr # SI-G1500
COMPARE
300 120 Orbital Shaking Incubator with Ratcheting Clamps, 120 VAC; US Standard Plug
$3,690.00USD / Each
Scientific Industries Genie Temp-Shaker 300 Orbital Shaking Incubator with Ratcheting Clamps, 230 VAC; British Plug EW-51603-24 Mfr # SI-G1503
COMPARE
300 230 Orbital Shaking Incubator with Ratcheting Clamps, 230 VAC; British Plug
$3,690.00USD / Each
Scientific Industries Genie Temp-Shaker 300 Orbital Shaking Incubator with Flask Clamp Platform, 120 VAC; US Standard Plug EW-51603-26 Mfr # SI-G1600
COMPARE
300 120 Orbital Shaking Incubator with Flask Clamp Platform, 120 VAC; US Standard Plug
$3,572.00USD / Each
Scientific Industries Genie Temp-Shaker 300 Orbital Shaking Incubator with Flask Clamp Platform, 230 VAC; British Plug EW-51603-29 Mfr # SI-G1603
COMPARE
300 230 Orbital Shaking Incubator with Flask Clamp Platform, 230 VAC; British Plug
$3,572.00USD / Each
Scientific Industries Genie Temp-Shaker 300 Orbital Shaking Incubator with Adhering Mat, 120 VAC; US Standard Plug EW-51603-31 Mfr # SI-G1700
COMPARE
300 120 Orbital Shaking Incubator with Adhering Mat, 120 VAC; US Standard Plug
$3,633.00USD / Each
Scientific Industries Genie Temp-Shaker 300 Orbital Shaking Incubator with Adhering Mat, 230 VAC; No Plug EW-51603-32 Mfr # SI-G1701
COMPARE
300 230 Orbital Shaking Incubator with Adhering Mat, 230 VAC; No Plug
$3,633.00USD / Each
Scientific Industries Genie Temp-Shaker 300 Orbital Shaking Incubator with Adhering Mat, 230 VAC; British Plug EW-51603-34 Mfr # SI-G1703
COMPARE
300 230 Orbital Shaking Incubator with Adhering Mat, 230 VAC; British Plug
$3,633.00USD / Each
Showing 1- 8 of 8