Scienceware Powder Funnels, PP

Use for transferring powders
  • 60°> wall incline
Polypropylene funnels have 60°> wall incline. Parallel stem minimizes bridging of powder.
$42.50 - $90.50USD / Pkg of 12

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stem OD (mm) Top OD (mm)
Availability Pricing
Scienceware H14660-0065 polypropylene powder funnel, 70 mL EW-06123-10 Mfr # H14660-0065
COMPARE
49 15 65
$56.80USD / Pkg of 12
Scienceware 14660-0080 polypropylene powder funnel, 125 mL EW-06123-20 Mfr # 14660-0080
COMPARE
138 18 80
$90.50USD / Pkg of 12
Scienceware 14660-0100 polypropylene powder funnel, 290 mL EW-06123-30 Mfr # 14660-0100
COMPARE
248 21 100
$42.50USD / Pkg of 6
Scienceware 14660-0150 polypropylene powder funnel, 700 mL EW-06123-40 Mfr # 14660-0150
COMPARE
784 29 150
$47.50USD / Pkg of 4
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat