Sartorius Entris® II Analytical Balances
This innovative balance features isoCAL, LED touch technology, and 12 built-in applications
  • Hybrid screen is easy to use and improves functionality
  • Optional isoCAL internal calibration saves you time and money for accurate weighing results
  • Parts offer a high chemical resistance so there is no need to worry over spills
  • Can transfer data directly to a PC with its optional data cable – no extra software required
Sartorius Entris® II analytical balances offer a better accuracy than their top-loading counterparts. Their draft shield protects samples from variance and provides more accurate weights. These models are perfect for laboratory use where they may encounter a variety of chemicals. The units are constructed of chemical resistant polybutylene terephthalate, stainless steel, and glass. In addition to accuracy and durability, the balances have advanced features that include dosing, counting, percentage weighing, mixing, totalizing, animal weighing, free factor, density determination, statistics, peakhold, checkweighing, mass unit conversion, and underfloor weighing. They are up to date with a USB type C interface for connecting to your PC. They are “PC-direct”, meaning that no extra software is required. You can download your data directly to Microsoft® Word or Excel.
ITEM(S) ON SALE
$2,103.20 - $2,956.80USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Power (VAC)
Availability Pricing
Sartorius BCE124-1S Entris® II Basic Essential Analytical Balance 120 g x 0.1 mg; 100 to 240 VAC, 50/60 Hz EW-11976-02 Mfr # BCE124-1S
COMPARE
External 120 100 to 240
$2,437.60USD / Each
$2,770.00USD / Each
Sartorius BCE124I-1S Entris® II Basic Essential Analytical Balance with Internal Calibration, 120 g x 0.1 mg; 100 to 240 VAC, 50/60 Hz EW-11976-03 Mfr # BCE124I-1S
COMPARE
Internal 120 100 to 240
$2,631.20USD / Each
$2,990.00USD / Each
Sartorius BCE224-1S Entris® II Basic Essential Analytical Balance 220 g x 0.1 mg; 100 to 240 VAC, 50/60 Hz EW-11976-10 Mfr # BCE224-1S
COMPARE
External 220 100 to 240
$2,754.40USD / Each
$3,130.00USD / Each
Sartorius BCE224I-1S Entris® II Basic Essential Analytical Balance with Internal Calibration, 220 g x 0.1 mg; 100 to 240 VAC, 50/60 Hz EW-11976-11 Mfr # BCE224I-1S
COMPARE
Internal 220 100 to 240
$2,956.80USD / Each
$3,360.00USD / Each
Sartorius BCE64-1S Entris® II Basic Essential Analytical Balance 60 g x 0.1 mg; 100 to 240 VAC, 50/60 Hz EW-11976-30 Mfr # BCE64-1S
COMPARE
External 60 100 to 240
$2,103.20USD / Each
$2,390.00USD / Each
Sartorius BCE64I-1S Entris® II Basic Essential Analytical Balance with Internal Calibration, 60 g x 0.1 mg; 100 to 240 VAC, 50/60 Hz EW-11976-31 Mfr # BCE64I-1S
COMPARE
Internal 60 100 to 240
$2,323.20USD / Each
$2,640.00USD / Each