Sartorius Cubis II (MCE) Ultra Micro and Micro Analytical Balances
Individual configuration of draft shield and integrated ionizer provides adaptation to any weighing task
  • Monolithical weighing system, integrated climate sensors, individual sample holders offer industry leading performance
  • Individual QApp workflows and motorized auto-leveling reduce error
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management provides excellent data integrity
  • Innovative TFT touch screen operation
  • Factory installed set of essential weighing applications
  • Essential user interface (MCE) features high-contrast touch display for easy operation
Ultra-Micro and Micro balances have a max capacity between 2.1 g and 10.1 g with a scale interval range of 0.1 μg to 1 μg. The automatic, motorized, round, glass draft shield has learning capability, illumination, and an integrated motion sensor. The manual stainless steel draft shield is best for filter weighing applications. Large, high-contrast touch display with factory-installed essential weighing applications for easy operation.
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Dual Range Capacity (g)
Availability Pricing
Sartorius Cubis II Ultra Micro Balance, 2.1 g x 0.0001 mg, with Manual Stainless Steel Draft Shield; 100 to 240 VAC, 50/60 Hz EW-39742-22 Mfr # MCE2.7S-2S00-F
COMPARE
Not Applicable 2.1
$40,154.40USD / Each
$45,630.00USD / Each
Sartorius Cubis II Ultra Micro Balance, 2.1 g x 0.0001 mg, with Automatic Glass Draft Shield; 100 to 240 VAC, 50/60 Hz EW-39742-23 Mfr # MCE2.7S-2S00-M
COMPARE
Not Applicable 2.1
$39,089.60USD / Each
$44,420.00USD / Each
Sartorius Cubis II Micro Balance, 6.1 g x 0.001 mg, with Automatic Glass Draft Shield; 100 to 240 VAC, 50/60 Hz EW-39742-24 Mfr # MCE6.6S-2S00-M
COMPARE
Not Applicable 6.1
$30,553.60USD / Each
$34,720.00USD / Each
Sartorius Cubis II Micro Poly Range Balance, 1.1|2.1|3.1 g x 0.001|0.002|0.005 mg, with Automatic Glass Draft Shield; 100 to 240 VAC, 50/60 Hz EW-39742-25 Mfr # MCE3.6PM-S00
COMPARE
1.1, 2.1, or 3.1 g 3.1
$23,364.00USD / Each
$26,550.00USD / Each