Sartorius® Cubis® II (MCA) Semi-Micro Analytical Balances with Software
Long-term flexibility for regulatory and laboratory needs
  • Fully customizable hardware, software, and connectivity
  • Complete data integrity for confident compliance
  • Motorized auto-leveling reduces errors
  • Error-free operation by assessing and correcting relevant factors
  • Electronic signature in compliance with 21 CFR Part 11
  • Safe data transfer
  • Advanced MCA user interface features large color TFT touch screen with swipe functionality
Sartorius® Cubis® II MCA semi micro balances are completely customizable to align with your unique application and compliance requirements while ensuring reliable results. They offer a max capacity between 60 g and 220 g with a scale interval range of 0.01 mg to 0.1 mg. Innovative technology includes automatic motorized leveling, motion controlled and illuminated draft shield, and climate sensor for environmental monitoring to ensure error-free operation.
Large 7” color touch-screen display allows you to touch, scroll, and swipe with factory-installed weighing applications. The status center shows all information about your balance and environmental conditions as well as detailed help and support in case of warnings and errors.
ITEM(S) ON SALE
$12,073.60 - $15,254.80USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Sartorius MCA225S-2S00-U Cubis® II Semi Micro Balance, 220 g x  0.01 mg; Manual Glass Draft Shield EW-39742-05 Mfr # MCA225S-2S00-U
COMPARE
220 0.01 Semi Micro Balance, 220 g x 0.01 mg; Manual Glass Draft Shield, w/o Software
$12,073.60USD / Each
$13,720.00USD / Each
Sartorius MCA225S-2S00-I Cubis® II Semi Micro Balance, 220 g x 0.01 mg; Auto Glass Draft Shield, Integrated Ionizer EW-39742-06 Mfr # MCA225S-2S00-I
COMPARE
220 0.01 Semi Micro Balance, 220 g x 0.01 mg; Auto Glass Draft Shield, Integrated Ionizer, w/o Software
$15,254.80USD / Each
$17,335.00USD / Each
Sartorius MCA225P-2S00-I Cubis® II Semi Micro Poly Range Balance, 60|120|220 g x 0.01|0.02|0.05 mg; Auto Glass Draft Shield, Integrated Ionizer EW-39742-07 Mfr # MCA225P-2S00-I
COMPARE
220 0.05 Semi Micro Poly Range Balance, 60|120|220 g x 0.01|0.02|0.05 mg; Auto Glass Draft Shield, Integrated Ionizer, w/o Software
$13,688.40USD / Each
$15,555.00USD / Each