SaniSure® Single-Use Sanitary Clamps, Nylon
Fast, safe, secure, and cost-effective connections!
  • Even 360-degree compression to prevent leakage
  • Validatable closure torque
  • Withstands pressures up to 100 psi
  • Reduces installation time
The SaniSure® clamps feature an innovative design for fast, safe, secure connections and offer a cost-effective solution for connecting single-use installations. Made of USP Class VI-approved nylon, these clamps are designed for single use applications but are durable enough to withstand repetitive use.
Unlike traditional two-point load stainless steel sanitary clamps, the design of these clamps ensure even 360-degree compression over the entire fitting surface—no more leaks and no gasket intrusion in your process line! Their low-profile rounded edges reduce the risk of penetrating bag assemblies, and the evenly controlled compression allows easy connection for plastic-to-plastic, plastic-to-stainless steel, or stainless steel-to-stainless steel coupling joints.
To use, simply insert the cable tie end until its made a couple clicks; position the clamp over the ferrule; hold both clamp parts together; move the clamp around the tri-clamp fitting joint and slide the clamp closed. Complete the closure by using the clamping tool (not included) to bring it to the proper tension.
The easy-to-use design reduces the chances of risk associated with carpal tunnel, or other wrist injuries, and yields a tamper-proof connection that is not operator-dependent. As a single-use device, they eliminate the need for cleaning validations and reduce your overall downtime for installation.
$26.75 - $28.75USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Connection Type Sanitary Clamp Size (Port 1) (in)
Availability Pricing
SaniSure SS-NYB-MINI Sanitary Clamp, Disposable; 3/4" Mini Connection Size EW-30562-21 Mfr # SS-NYB-MINI
COMPARE
Nylon Sanitary 3/4
$26.75USD / Each
SaniSure SS-NYB-150 Sanitary Clamp, Disposable; 1-1/2" Connection Size EW-30562-22 Mfr # SS-NYB-150
COMPARE
Nylon Sanitary 1-1/2
$28.75USD / Each