Scienceware Sampler Syringe, Polypropylene
Invert syringe to contain sample in the graduated bulb
  • Constructed with polypropylene syringe and polyethylene squeeze bulb
  • Reusable syringe can be unscrewed from bulb for easy cleaning and storage
Sample liquids quickly and easily - simply squeeze the bulb, submerse the syringe in liquid, and then release the bulb to draw liquids into the syringe. Either transfer the liquid to another vessel or invert the syringe and the sample is in the graduated bulb. Syringe features a graduated capacity of 30 mL (1 oz) and the squeeze bulb has a 100 mL capacity with 5 mL graduations. This sampler syringe handles liquids up to 80° C (176°F) and has an over length of 11.75” (30 cm).
$33.80 - $366.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Syringe Capacity (mL) Description
Availability Pricing
Scienceware Sampler Syringe, Polypropylene, 100 mL; 1/Ea EW-07954-10 Mfr # 378790000
COMPARE
Polypropylene syringe and polyethylene bulb 100 Sampler Syringe, Polypropylene, 100 mL; 1/Ea
$33.80USD / Each
Scienceware Sampler Syringe, Polypropylene, 100 mL; 12/Pk EW-07954-20 Mfr # 378790000
COMPARE
Polypropylene syringe and polyethylene bulb 100 Sampler Syringe, Polypropylene, 100 mL; 12/Pk
$366.00USD / Pkg of 12