Salisbury by Honeywell Class 0 Insulated Rubber Gloves
Protect yourself when working with switchboards, power cabinets, or anytime you are working on energized circuits. Combining high dielectric and physical strength, flexibility and durability, gloves provide superior performance meeting and exceeding the requirements of current ASTM D120 specifications and NFPA 70E standards.
* Made of Type I natural rubber
* Meet or exceed ASTM D120 and IEC EN60903
* Available in different colors to allow easy inspection of a glove rotation program
* Glove flexibility makes working with small parts easy
* Use Glove dust (sold separately) to absorb moisture and perspiration when wearing gloves
* Glove Inflator (sold separately) simplifies inspection of gloves before each use
$123.25 - $137.00USD / Pair of 2
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Size
Availability Pricing
Salisbury by Honeywell E011R8H Insulated Glove, Class 0, Red, 11" length, size 8-1/2 EW-86432-22 Mfr # E011R8H
COMPARE
Red 11 8.5
$123.25USD / Pair of 2
Salisbury by Honeywell E011B9 Insulated Glove, Class 0, Black, 11" length, size 9 EW-86432-33 Mfr # E011B9
COMPARE
Black 11 9
$123.25USD / Pair of 2
Salisbury by Honeywell E011B10 Insulated Glove, Class 0, Black, 11" length, size 10 EW-86432-35 Mfr # E011B/10
COMPARE
Black 11 10
$123.25USD / Pair of 2
Salisbury by Honeywell E011B11 Insulated Glove, Class 0, Black, 11" length, size 11 EW-86432-37 Mfr # E011B/11
COMPARE
Black 11 11
$123.25USD / Pair of 2
Salisbury by Honeywell E011Y8 Insulated Glove, Class 0, Yellow, 11" length, size 8 EW-86432-41 Mfr # E011Y/8
COMPARE
Yellow 11 8
$137.00USD / Pair of 2
Salisbury by Honeywell E011Y9 Insulated Glove, Class 0, Yellow, 11" length, size 9 EW-86432-43 Mfr # E011Y/9
COMPARE
Yellow 11 9
$137.00USD / Pair of 2
Salisbury by Honeywell E011Y10 Insulated Glove, Class 0, Yellow, 11" length, size 10 EW-86432-45 Mfr # E011Y/10
COMPARE
Yellow 11 10
$137.00USD / Pair of 2
Showing 1- 7 of 7