Rubberfab Torque Tee Sanitary Clamp Securing Devices
Ensure the security of sanitary connections with preset torque output
  • Works with any sanitary clamp wing nut and Rubberfab's Torque-Rite® nut
  • Easily reach into tight or awkward spaces
  • Choose the appropriate torque output per the clamp manufacturer's specifications
  • Audible click alerts you when the torque has been reached
  • Ratcheting mechanism disengages for easy clamp removal and is sealed within handle for durability
  • "T" shape creates effective leverage for fast, easy one-handed tightening
The Rubberfab Torque Tee and universal socket are designed to safely and securely install and remove sanitary clamps. Available with a family of components, the Torque Tee handle creates effective tightening leverage, even in hard-to-reach spaces. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle. The universal socket is required when using the Torque Tee.
$40.75 - $213.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Rubberfab TR-50-TEE-CP Torque Tee Sanitary Clamp Securing Handle, 50 in-lb EW-23508-03 Mfr # TR-50-TEE-CP
COMPARE
Torque Tee Sanitary Clamp Securing Handle, 50 in-lb
$213.00USD / Each
Rubberfab Torque Tee Sanitary Clamp Securing Device, 3-Piece Extension Set, 1/4" EW-23508-04 Mfr # TR-EXT-.25-SET
COMPARE
Torque Tee Sanitary Clamp Securing Device, 3-Piece Extension Set, 1/4"
$40.75USD / Each
Rubberfab Torque Tee Sanitary Clamp Securing Device, Swivel Joint, 1/4" EW-23508-05 Mfr # TR-SWIVEL-.25
COMPARE
Torque Tee Sanitary Clamp Securing Device, Swivel Joint, 1/4"
$136.00USD / Each
Rubberfab TR-PW Torque Tee Sanitary Clamp Securing Device, Palm Wrench, 3/8" EW-23508-06 Mfr # TR-PW
COMPARE
Torque Tee Sanitary Clamp Securing Device, Palm Wrench, 3/8"
$54.00USD / Each
Rubberfab TR-PW-.25 Torque Tee Sanitary Clamp Securing Device, Palm Wrench, 1/4" EW-23508-07 Mfr # TR-PW-.25
COMPARE
Torque Tee Sanitary Clamp Securing Device, Palm Wrench, 1/4"
$54.00USD / Each
Rubberfab Torque Tee Sanitary Clamp Securing Device, Universal Socket
Always in Stock
EW-23508-08 Mfr # WN/TR SOCKET
COMPARE
Torque Tee Sanitary Clamp Securing Device, Universal Socket
$140.00USD / Each
Rubberfab TR-50 Torque-Rite® Nut, 50 in-lb; 1/Each EW-38450-85 Mfr # TR-50
COMPARE
Torque-Rite® Nut, 50 in-lb
$158.00USD / Each
Showing 1- 7 of 7