Rotator Genie Rotating Shakers
Designed for a wide range of motions from gentle rolling to forceful end-over-end mixing
  • Hold a variety of tubes, flasks, microplates, and other vessels at any angle
  • Available with a standard or magnetic platform which are easily interchangeable
  • Cold room safe and easily fits in most incubators
  • Motor provides smooth, continuous, and quiet operation
Select from the standard platform which can be turned 90 degrees to allow for different mixing motions and includes retaining bands, or the magnetic stainless steel platform which features a spring clip plate system (plates sold separately).
$1,115.00 - $1,156.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Degree of Rotation (degrees) Power (VAC) Description
Availability Pricing
Scientific Industries Genie SI-2100 Rotator with Standard Platform; 120V EW-51603-11 Mfr # SI-2100
COMPARE
360 120 Rotator with Standard Platform; 120V
$1,156.00USD / Each
Scientific Industries Genie SI-2101 Rotator with Standard Platform; 230V EW-51603-12 Mfr # SI-2101
COMPARE
360 230 Rotator with Standard Platform; 230V
$1,115.00USD / Each
Scientific Industries Genie SI-2201 Rotator with Magnetic Platform; 230V EW-51603-14 Mfr # SI-2201
COMPARE
360 230 Rotator with Magnetic Platform; 230V
$1,151.00USD / Each