InnoCal® Rotational Viscometer Calibration
Ensure your instrument remains precise
  • Receive your product calibrated and ready to use
  • Confirm that your item meets the manufacturer's published specification
  • Meet EPA, USDA, ISO, and GLP/GMP standards
  • Order A2LA endorsement separately
Rotational viscometers are tested at the midpoint value of the spindles. Accuracy is confirmed using ISO Guide 34 Accredited Standard Reference Materials.
$120.50 - $416.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Number Of Calibration Test Points
Availability Pricing
COMPARE
Liquid Testing 1 Per Spindle
$343.00USD / Each
COMPARE
$120.50USD / Each
COMPARE
A2LA ISO17025 Accredited Liquid Testing 1 Per Spindle
$416.00USD / Each