Reichert Rhino Series Brix Refractometers
Feature automatic temperature compensation—no correction tables or temperature measurement required
  • Water-resistant IP67-rated design is ideal for stringent environments
  • Rugged, impact-resistant Xenoy® polycarbonate housing withstands repeated drops
These refractometers offer exceptional optical design and improved image resolution with high contrast for easy viewing of scales. All reflection-coated glass optical components, including a 7x eyepiece, have been developed to improve illumination and shadowline contrast. Black housing improves image quality by reducing stray light. Optics and instrument scales are sealed inside, safe from harsh external conditions. Measure concentration of fruits, beverages, and canned goods; total solid content of syrups, vinegar, and sauces; or check the quality of vegetable, fruit, and grass tissues.
$275.00 - $280.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Brix Range (%) Resolution
Availability Pricing
Reichert 137530LO Handheld Brix Refractometer; 0 to 30% EW-81007-50 Mfr # 137530L0
COMPARE
30 0.2% (Brix)
$275.00USD / Each
Reichert 137531LO Handheld Brix Refractometer; 0 to 50% EW-81007-52 Mfr # 137531LO
COMPARE
50 0.25% (Brix)
$275.00USD / Each
Reichert 137533LO Handheld Brix Refractometer; 50 to 90% EW-81007-54 Mfr # 137533L0
COMPARE
90 0.2% (Brix)
$280.00USD / Each