This product is now discontinued, View Related Products
Red Lion Modular Controllers
Red Lion’s Modular Controller series offers a cost effective solution to integrating multi-zone PID control, data acquisition and I/O into your PC, DCS or PLC control system. Users drag and drop controller data to PLC registers in seconds using point-and-click data mapping from an extensive menu of built-in serial and Ethernet drivers. There is no tedious PLC code to write. Instantly Ethernet-enable your PLC for free! Not only are programming costs slashed, you can eliminate costly add-on I/O and analog cards and solve applications simply using an inexpensive brick PLC. The high-density DIN rail design saves panel space and eliminates cutouts. All modules can be hot-swapped - the master automatically programs a replaced module.
Call For Price
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Red Lion CSDIO14R Modular Controller 8 Inputs 6 Relay Out EW-68483-93 Mfr # CSDIO14R
COMPARE
Modular Controller 8 Inputs 6 Relay Out
Call For Price
Red Lion CSDIO14S Modular Controller 8 Inputs 6 Solid State Outputs EW-68483-94 Mfr # CSDIO14S
COMPARE
Modular Controller 8 Inputs 6 Solid State Outputs
Call For Price
Red Lion CSINI800 8 Cannel 0 (4) 20 Ma Input Module EW-68483-95 Mfr # CSINI800
COMPARE
8 Cannel 0 (4) 20 Ma Input Module
Call For Price
Red Lion CSINV800 Modular Controller 8 Ch +/- 10V Input EW-68483-96 Mfr # CSINV800
COMPARE
Modular Controller 8 Ch +/- 10V Input
Call For Price
Red Lion CSMSTRSE Controller Series Master Comms Ethernet EW-68483-97 Mfr # CSMSTRSE
COMPARE
Controller Series Master Comms Ethernet
Call For Price
Red Lion CSPID1R0 Modular Controller Single Loop Relay Outs EW-68483-98 Mfr # CSPID1R0
COMPARE
Modular Controller Single Loop Relay Outs
Call For Price
Red Lion CSPID1RA Modular Controller Single Loop Relay Outs Analog EW-68483-99 Mfr # CSPID1RA
COMPARE
Modular Controller Single Loop Relay Outs Analog
Call For Price
Red Lion CSPID1RM Modular Controller Single Loop Relay Outs Hcm EW-68484-00 Mfr # CSPID1RM
COMPARE
Modular Controller Single Loop Relay Outs Hcm
Call For Price
Red Lion CSPID1SA Modular Controller Single Loop Relay Outs Analog EW-68484-01 Mfr # CSPID1SA
COMPARE
Modular Controller Single Loop Relay Outs Analog
Call For Price
Red Lion CSPID1SM Modular Controller Single Loop Relay Outs Hcm EW-68484-02 Mfr # CSPID1SM
COMPARE
Modular Controller Single Loop Relay Outs Hcm
Call For Price
Red Lion CSPID1S0 CSPID1SO:Modular Controller Single Loop Solid State Outs EW-68484-03 Mfr # CSPID1S0
COMPARE
CSPID1SO:Modular Controller Single Loop Solid State Outs
Call For Price
Red Lion CSPID1TA Modular Controller Single Loop Triac Outs Analog EW-68484-04 Mfr # CSPID1TA
COMPARE
Modular Controller Single Loop Triac Outs Analog
Call For Price
Red Lion CSPID2RM Modular Controller Dual Loop Relay Outs Hcm EW-68484-06 Mfr # CSPID2RM
COMPARE
Modular Controller Dual Loop Relay Outs Hcm
Call For Price
Red Lion CSPID2SM Dual Loop Module Solid State Outputs Hcm EW-68484-08 Mfr # CSPID2SM
COMPARE
Dual Loop Module Solid State Outputs Hcm
Call For Price
Red Lion CSPID2TM Modular Controller Dual Loop Triac Outs Hcm EW-68484-10 Mfr # CSPID2TM
COMPARE
Modular Controller Dual Loop Triac Outs Hcm
Call For Price
Red Lion CSRTD600 Modular Controller, 6-Channel RTD EW-68484-11 Mfr # CSRTD600
COMPARE
Modular Controller, 6-Channel RTD
Call For Price
Red Lion CSTC8000 Modular Controller, 8-Channel Thermocouple EW-68484-12 Mfr # CSTC8000
COMPARE
Modular Controller, 8-Channel Thermocouple
Call For Price
Showing 1- 17 of 17