Reagents® Lithium Chloride, Reagent

Manufactured using the highest quality raw materials in ISO 9001 facility
  • Minimum Assay 99%
Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
$145.75 - $2,110.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Class Weight (g) Description
Availability Pricing
Reagents Lithium Chloride, 100g, Reagent EW-78911-86 Mfr # C1233500-100C2
COMPARE
Salts 100 Lithium Chloride, 100g, Reagent
$145.75USD / Each
Reagents Lithium Chloride, 500g, Reagent EW-78918-80 Mfr # C1233500-500C2
COMPARE
Salts 500 Lithium Chloride, 500g, Reagent
$548.50USD / Each
Reagents Lithium Chloride, 2.5kg, Reagent EW-78919-18 Mfr # C1233500-2.5A5
COMPARE
Salts 2500 Lithium Chloride, 2.5kg, Reagent
$2,110.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat