Radiation Alert® Personal Pen Dosimeters
Direct-reading dosimeter measures gamma and x-ray radiation exposure
  • Rugged charger requires no batteries — simply squeeze the trigger of this piezoelectric generator to move the hairline fiber inside the dosimeter back to zero
  • Clamping mechanism automatically locks the dosimeter in place
  • Charger is easy to operate and requires only few squeezes of the trigger for each dosimeter
Personal, pen-style dosimeters clip easily to your pocket or a specific object to measure personal accumulated dose or quantity of gamma and x-ray radiation. Dosimeter is a conductive-fiber electroscope with a thin wall to allow penetration of radiation. Inside the dosimeter, the fiber moves across a predetermined scale to display the level of radiation exposure. Readings are easily displayed by holding the dosimeter up to a light source to view the scale — immediate results without external lab analysis. Order handheld charger (sold separately) to zero dosimeters and reset scale for repeated use.
$203.75 - $204.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Range
Availability Pricing
Radiation Alert Personal Radiation Dosimeter, 0 to 200 Mr EW-81922-10 Mfr # PEN200MR
COMPARE
0 to 200 mR
$203.75USD / Each
Radiation Alert Personal Radiation Dosimeter, 0.1 to 2.0 Msv EW-81922-15 Mfr # PEN2MSV
COMPARE
0.1 to 2 mSv
$204.00USD / Each