R M Young Wind Monitor Transmitter and Indicator
The Wind Monitor is a high resolution rugged wind sensor
  • Lightweight
  • Corrosion resistant
Its simplicity and lightweight corrosion resistant construction make it well suited for a wide range of wind measuring applications. The wind speed sensor is a four blade helicoid propeller. Propeller rotation produces an AC sine wave voltage signal with frequency directly proportional to wind speed. Slip rings and brushes are not used resulting in improved reliability and lower cost. The wind direction sensor is a rugged yet lightweight vane with a sufficiently low aspect ratio to assure good fidelity in fluctuating wind conditions. Vane position is sensed by a precision conductive plastic potentiometer located in a sealed chamber.
$1,584.00 - $2,255.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
R. M. Young 6201 Wind Tracker Indicator with Alarms EW-99536-17 Mfr # 06201
COMPARE
Wind Tracker Indicator with Alarms
$1,584.00USD / Each
R. M. Young 5103 Wind Monitor Sensor, wind speed and direction EW-99536-18 Mfr # 05103
COMPARE
Wind Monitor Sensor, wind speed and direction
$1,686.00USD / Each
R. M. Young 05103L Wind Monitor Sensor, 4-20 mA output EW-99536-19 Mfr # 05103L
COMPARE
Wind Monitor Sensor, 4-20 mA output
$2,255.00USD / Each