Quartz Cuvettes for Fluorometry
Matched pairs for less data variability
  • Matched to give the same value for the same solution within 1% solerance
  • Guaranteed 80% transmission at 200 nm
Cuvettes come in matched pairs in standard, semimicro, or micro size, and can be ordered with either a cover of PTFE resin or a stopper of PTFE resin. All cuvettes have a 10-mm pathlength. All pairs are accurately matched to give the same value for the same solution within 1% tolerance. Quartz cuvettes can be used for the entire UV spectra down to 200 nm and have a guaranteed 80% transmission at 200 nm.
$552.00USD / Pkg of 2
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (mm) Max Capacity (mL) Pathlength (mm)
Availability Pricing
COMPARE
45 3.5 10
$552.00USD / Pkg of 2