PTFE Thread Tape (Plumber's)
PTFE Plumber's Tape is used on all types of threaded fittings for passage of potable and non-potable water, most chemicals, oxygen, gas, hydraulic and high-pressure lines. Withstands harsh temperatures from -450 to 550°F (-267 to 287°C). Strong chemical resistance. Stretches 50% without breaking.
$8.40 - $8.60USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Thickness (mil) Width (in)
Availability Pricing
COMPARE
43 3.5 1/2
$8.40USD / Each
COMPARE
43 3.5 3/4
$8.60USD / Each