Powder Pack and Tube Test Reagents
Premeasured reagents in convenient powder packs
  • Precise dosing with simple push-button operation
These powder packs contain premeasured unit dose reagents suitable for use with Lovibond/Orbeco-Hellige colorimeters, Aqua comparator test kits, YSI, Thermo, and Hach® instrumentation/test kits. Reagents for chlorine are DPD chemistry. Some parameters require multiple reagents and are bundled into a "Reagent set", which can include powder packs, tube tests, or both.
$29.00 - $261.00USD / Pkg of 1
22 variations of this product are available.
See All Product Options Below
22 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Tests Included Test Parameters
Availability Pricing
Lovibond RP009-0 Powder Pillow, DPD Free Chlorine Reagent, 5 mL, 100 tests EW-99560-52 Mfr # 530090
COMPARE
100 x 5 mL DPD free chlorine
$29.80USD / Pkg of 1
Lovibond RP009-3 Powder Pillow, DPD Free Chlorine Reagent, 5 mL, 1000 tests EW-99560-53 Mfr # 530093
COMPARE
1000 x 5 mL DPD free chlorine
$209.00USD / Pkg of 1
Lovibond RP010-0 Powder Pillow, DPD Free Chlorine Reagent, 10 mL, 100 tests
Always in Stock
EW-99560-54 Mfr # 530100
COMPARE
100 x 10 mL DPD free chlorine
$29.00USD / Pkg of 1
Lovibond RP010-3 Powder Pillow, DPD Free Chlorine Reagent, 10 mL, 1000 tests EW-99560-55 Mfr # 530103
COMPARE
1000 x 10 mL DPD free chlorine
$245.00USD / Pkg of 1
Lovibond RP011-0 Powder Pillow, DPD Free Chlorine Reagent, 25 mL, 100 tests EW-99560-56 Mfr # 530110
COMPARE
100 x 25 mL DPD free chlorine
$29.30USD / Pkg of 1
Lovibond RP011-3 Powder Pillow, DPD Free Chlorine Reagent, 25 mL, 1000 tests EW-99560-57 Mfr # 530113
COMPARE
1000 x 25 mL DPD free chlorine
$249.00USD / Pkg of 1
Lovibond RP012-0 Powder Pillow, DPD Total Chlorine Reagent, 10 mL, 100 tests EW-99560-58 Mfr # 530120
COMPARE
100 x 10 mL DPD total chlorine
$29.00USD / Pkg of 1
Lovibond RP012-3 Powder Pillow, DPD Total Chlorine Reagent, 10 mL, 1000 tests EW-99560-59 Mfr # 530123
COMPARE
1000 x 10 mL DPD total chlorine
$261.00USD / Pkg of 1
Lovibond RP013-0 Powder Pillow, DPD Total Chlorine Reagent, 25 mL, 100 tests EW-99560-60 Mfr # 530130
COMPARE
100 x 25 mL DPD total chlorine
$29.50USD / Pkg of 1
Lovibond RP013-3 Powder Pillow, DPD Total Chlorine Reagent, 25 mL, 1000 tests EW-99560-61 Mfr # 530133
COMPARE
1000 x 25 mL DPD total chlorine
$251.00USD / Pkg of 1
Lovibond RP030-0 Powder Pillow, Copper Reagent, 10 mL, 100 tests EW-99560-62 Mfr # 530300
COMPARE
100 x 10 mL Copper
$60.50USD / Pkg of 1
Lovibond RP055-0 Powder Pillow, Iron TPTZ Reagent, 10 mL, 100 tests EW-99560-63 Mfr # 530550
COMPARE
100 x 10 mL Iron TPTZ
$44.30USD / Pkg of 1
Lovibond RP056-0 Powder Pillow, Iron Ferro Reagent, 10 mL, 100 tests EW-99560-64 Mfr # 530560
COMPARE
100 x 10 mL Iron ferro
$30.30USD / Pkg of 1
Lovibond RS5100 Powder Pillow, Manganese Reagent, High Range, 10 mL, 100 tests EW-99560-66 Mfr # 535100
COMPARE
100 x 10 mL Manganese, high range
$83.80USD / Pkg of 1
Lovibond RS5580 Tube Test Set, Nitrate Reagent, 50 tests EW-99560-69 Mfr # 535580
COMPARE
50 Nitrate
$108.00USD / Pkg of 50
Lovibond RP098-0 Powder Pillow, Nitrite, 10 mL, 100 tests EW-99560-71 Mfr # 530980
COMPARE
100 x 10 mL Nitrite
$51.00USD / Pkg of 1
Lovibond RS5650 Colorimeter Reagent Set, Ammonia, High Range, 50 tests EW-99560-73 Mfr # 535650
COMPARE
50 Ammonia, high range
$130.00USD / Pkg of 1
Lovibond RS5550 Colorimeter Reagent Set, Total Nitrogen, Low Range, 50 tests EW-99560-75 Mfr # 535550
COMPARE
50 Total nitrogen, low range
$188.00USD / Pkg of 1
Lovibond RP155-0 Powder Pillow, Phosphate 3, 10 mL, 100 tests EW-99560-77 Mfr # 531550
COMPARE
100 x 10 mL Phosphate
$44.30USD / Pkg of 1
Lovibond RS5210 Tube Test Set, Total Phosphate Reagent, 50 tests EW-99560-79 Mfr # 535210
COMPARE
50 Total phosphate
$87.50USD / Pkg of 50
Showing 1 - 20 of 22