Cole-Parmer® Essentials Polystyrene Weighing Dishes - Vessels (Anti-Static)
Excellent for weighing static-affected materials
  • Disposable
Translucent polystyrene flexes easily for weighing, dispensing, or pouring liquids or solids. Unique design facilitates non-slip handling and pouring. Biologically inert; resistant to most dilute acids, aqueous solutions, and alcohol bases. Flat bottoms resist tipping. Three convenient sizes for any sampling or weighing applications.
$46.00 - $132.00USD / Pkg of 500
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Length (in) Width (in)
Availability Pricing
COMPARE
20 1 1/2 2 1/8
$46.00USD / Pkg of 500
COMPARE
120 3 1/4 5 1/4
$67.00USD / Case of 500
COMPARE
240 7 1/4 7
$132.00USD / Case of 500