PolyScience Constant-Temperature Viscosity Baths
An economical alternative for kinematic viscosity measurement
These baths are ideal for measuring petroleum, lubricants, polymer coatings, food, paint, and other liquids' kinematic viscosity. Advanced PID controls maintain precise temperatures and quick responsiveness under changing heat load conditions. The integral recirculating pump ensures uniform temperature. Large, backlit digital display shows both set and actual values. Other features include Celsius/Fahrenheit selection, high/low temperature limits, chemical- and thermal-resistant top plate, and transparent tank for excellent visibility.
$3,828.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Reservoir Capacity (L)
Availability Pricing
PolyScience MX17VB5G-A11 Constant-Temperature Viscosity Baths, 17 L, 115 VAC EW-98935-22 Mfr # MX17VB6G-A11B
COMPARE
115 17
$3,828.00USD / Each
PolyScience MX17VB5G-A12 Constant-Temperature Viscosity Bath, 17 L, 240 VAC EW-98935-24 Mfr # MX17VB6G-A12E
COMPARE
240 17
$3,828.00USD / Each