Polypropylene (PP) High-Head Chemical-Resistance Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
High-head polypropylene pumps have a closed impeller design to significantly increase the output pressure. Polypropylene construction provides excellent chemical resistance in applications, such as filtration, where a high-head is needed.
$370.90 - $731.75USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Power (VAC)
Availability Pricing
COMPARE
2.9 22.6 115
$380.00USD / Each
COMPARE
2.9 22.6 230
$370.90USD / Each
COMPARE
4.6 36.1 115
$420.30USD / Each
COMPARE
4.6 36.1 230
$427.60USD / Each
COMPARE
11.4 66.6 115
$702.25USD / Each
COMPARE
11.4 66.6 230
$731.75USD / Each