Cole-Parmer® Essentials Platinum Inoculation Loops and Needles
Ideal for milk and bacterial cultures
Calibrated loops 01850-00 and 01850-02 are ideal for milk samples. The 1-microliter loops are made of 26-ga B&S platinum wire with 15% iridium. The 10-microliter loops are calibrated and made of 19-ga B&S platinum wire with 5% rhodium. Order insulated loop and needle holder separately.
Platinum loops and needles are used for transferring bacterial cultlres. These loops and needles are made of B&S platinum wire with 15% iridium added for rigidity, permitting a thinner wire for delicate work. Loops are fused to form a "perfect circle" with an inside tolerance of +/-0.02 mm to ensure accurate delivery. Order insulated loop and needle holder separetly.
$199.00 - $260.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
B and S Wire Gauge Loop Length (in) Description
Availability Pricing
COMPARE
26 2 3/4 Calibrated loop, platinum, 1 microliter, 1.45 mm ID, 70 mm length
$203.00USD / Each
COMPARE
19 1 Calibrated loop, platinum, 10 microliter, 4 mm ID, 25 mm length
$260.00USD / Each
COMPARE
24 1 3/4 Inoculating loop, platinum, 2 mm ID, 24 gauge, 45 mm length
$199.00USD / Each
COMPARE
24 1 3/4 Inoculating loop, platinum, 3 mm ID, 24 gauge, 45 mm length
$203.00USD / Each
COMPARE
24 1 3/4 Inoculating loop, platinum, 5 mm ID, 24 gauge, 45 mm length
$218.00USD / Each
COMPARE
26 1 3/4 Inoculating loop, platinum, 3 mm ID, 26 gauge, 45 mm length
$207.00USD / Each
COMPARE
26 2 3/4 Inoculating loop, platinum, 3 mm ID, 26 gauge, 70 mm length
$206.00USD / Each
COMPARE
26 2 3/4 Inoculating needle, 26 gauge, 70 mm length
$216.00USD / Each
Showing 1- 8 of 8