pHydrion Sanitizer Strength Test Strips
Convenient dispenser roll lets you use the exact strip length needed
  • Reliable and economical means to measure sanitizer strength
Federal, state, and local health regulations require the use of test kits to verify the strength of sanitizer solutions. Hydrion test strips are a quick, reliable, and economical solution for this. Test strips are available to test quaternary ammonium, chlorine, and iodine-based sanitizers. Strips provide a visual color-change indication of solution strength. Each 15-ft (4.6-m) roll comes with a color match chart and is clearly labeled. Each roll provides enough paper for about 100 tests.
$75.60 - $87.70USD / Box of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test
Availability Pricing
Hydrion QT-10 quaternary ammonium test paper, 15-ft roll, box of 10
Always in Stock
EW-59200-50 Mfr # QT-10
COMPARE
0, 100, 200, 300, 400 ppm Quaternary ammonium
$86.10USD / Box of 10
Hydrion CM-240 chlorine test paper, 15-ft roll, box of 10
Always in Stock
EW-59200-52 Mfr # CM-240
COMPARE
10, 50, 100, 200 ppm Chlorine
$75.60USD / Box of 10
Hydrion IM-220 iodine test paper, 15-ft roll, box of 10 EW-59200-54 Mfr # IM-220
COMPARE
50, 75, 150, 225 ppm Iodine
$87.70USD / Box of 10