pHydrion Plastic pH Strips in Flip Top Vials
Three times the matching area compared to other plastic strips
  • Simple, single color match
  • Handy matching color chart
  • Long lasting colors
Immediate pH testing is available in brilliant, long lasting colors. These direct-reading pH papers give a simple, single color match for all pH values in distinct, bright color separations. Handy flip top vials contain 100 dip sticks and feature a handy matching color chart.
$25.60 - $30.00USD / Pkg of 100
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Tests Per Kit
Availability Pricing
Hydrion 9800 plastic pH indicator strips, 0.0 to 14.0, flip top vial packaging
Always in Stock
EW-59200-40 Mfr # 9800
COMPARE
0 to 14 pH in 1 pH unit increments 100
$25.60USD / Pkg of 100
Hydrion 9200 Plastic pH indicator strips, 0.0 to 6.0, flip top vial packaging
Always in Stock
EW-59200-42 Mfr # 9200
COMPARE
0 to 6 pH in 0.5 pH unit increments 100
$29.80USD / Pkg of 100
Hydrion 9400 Plastic pH indicator strips, 5.0 to 9.0, flip top vial packaging
Always in Stock
EW-59200-44 Mfr # 9400
COMPARE
5 to 9 pH in 0.5 pH unit increments 100
$29.80USD / Pkg of 100
Hydrion 9600 Plastic pH indicator strips, 6.5 to 13.0, flip top vial packaging EW-59200-46 Mfr # 9600
COMPARE
6.5 to 13 pH in 0.5 pH unit increments 100
$30.00USD / Pkg of 100
Hydrion 9700 Plastic pH indicator strips, 5.5 to 8.0, flip top vial packaging
Always in Stock
EW-59200-48 Mfr # 9700
COMPARE
5.5 to 8 pH in 0.5 pH unit increments 100
$29.80USD / Pkg of 100