pHydrion Jumbo pH Papers
Larger, field-sized papers for quick pH monitoring
These low-cost jumbo rolls offer larger paper strips to measure pH in one of four ranges. Ideal for use in the field or plant. The larger size makes it easier for testing--even with gloves.
$22.60 - $25.20USD / Carton of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Points Test Description
Availability Pricing
Hydrion pH 1 to 12 Jumbo pH Papers, Range 1 to 12, 50 Ft/Roll
Always in Stock
EW-59200-22 Mfr # HJ-600
COMPARE
1 to 12 pH in 1 pH unit increments pH Jumbo pH Papers, Range 1 to 12, 50 Ft/Roll
$24.50USD / Carton of 1
Hydrion 0 to 13 pH Jumbo pH Papers, Range 0 to 13, 50 Ft/Roll
Always in Stock
EW-59200-24 Mfr # HJ-613
COMPARE
1 to 13 pH in 1 pH unit increments pH Jumbo pH Papers, Range 0 to 13, 50 Ft/Roll
$22.60USD / Carton of 1
Hydrion 3.0 to 5.5 pH Jumbo pH Papers, Range 3.0 to 5.5, 50 Ft/Roll EW-59200-26 Mfr # HJ-626
COMPARE
3.0 to 5.5 pH in 0.5 pH unit increments pH Jumbo pH Papers, Range 3.0 to 5.5, 50 Ft/Roll
$25.00USD / Carton of 1
Hydrion 6.0 to 8.0 pH Jumbo pH Papers, Range 6.0 to 8.0, 50 Ft/Roll EW-59200-28 Mfr # HJ-633
COMPARE
6 to 8 pH pH Jumbo pH Papers, Range 6.0 to 8.0, 50 Ft/Roll
$25.20USD / Carton of 1