PendoTECH® Single-Use Pressure Sensors for Pressure Monitor and Transmitter Systems
Single-use solution to monitor process pressure
  • Ideal for applications such as tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring, and chromatography
  • Sensors priced for single-use systems, but may be reused
Sensors measure static and dynamic process pressure and are perfect components for disposable systems requiring sterilization, but also minimal downtime. Sensors feature an integral flow sensor with no dead legs and no fluid path obstructions. All pressure sensors are manufactured in a FDA compliant facility, in an ISO Class 8 clean room. Sensors are made from USP Class VI-compliant polycarbonate or caustic-resistant polysulfone. Both materials are gamma stable. Sensors are 100% tested for accuracy and leaks during the manufacturing process. Connect sensor to PressureMAT unit with 12-ft (3.6-m) reusable cable (included). Sensors are designed for use up to a maximum pressure of 5.3 bar. Available in either process–scale or small–scale sizes.
Process-Scale Sensors connect directly as a barbed fitting to the tubing line and are available in 1⁄4", 3⁄8", 1⁄2", 3⁄4", or 1" hose barb or 3/4" or 1" sanitary flange.
Small-Scale Sensors are available with luer connections and may connect directly to the fluid line with luer adapters or indirectly by using a 1⁄4" or 3⁄8" barbed adapter tee. Both connection setups have been tested for accuracy.
$54.00 - $121.00USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Process Connection Wetted Materials
Availability Pricing
PendoTech PREPS-N-000 Single-Use Pressure Sensor, Non-Sterile, Polysulfone, Luer Fitting EW-19406-20 Mfr # PREPS-N-000
COMPARE
75 Female Luer Lock Polysulfone
$67.00USD / Each
PendoTech PRESS-N-025 Single-Use Pressure Sensor, Non-Sterile, Polycarbonate, 1/4" Hose Barb
Best Seller Products
EW-19406-21 Mfr # PRESS-N-025
COMPARE
75 1/4" Hose Barb Polycarbonate
$75.00USD / Each
PendoTech PREPS-N-050 Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 1/2" Hose Barb EW-19406-22 Mfr # PREPS-N-050
COMPARE
75 1/2" Hose Barb Polysulfone
$103.00USD / Each
PendoTech PRESS-N-038 Single-Use Pressure Sensor, Non-Sterile, Polycarbonate, 3/8" Hose Barb EW-19406-23 Mfr # PRESS-N-038
COMPARE
75 3/8" Hose Barb Polycarbonate
$78.00USD / Each
PendoTech PREPS-N-075 Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 3/4" Hose Barb EW-19406-24 Mfr # PREPS-N-075
COMPARE
75 3/4" Hose Barb Polysulfone
$107.00USD / Each
PendoTech PREPS-N-038 Single-Use Pressure Sensor, Non-Sterile, Polysulfone 3/8" Hose Barb EW-19406-25 Mfr # PREPS-N-038
COMPARE
75 3/8" Hose Barb Polysulfone
$93.00USD / Each
PendoTech PREPS-N-025 Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 1/4" Hose Barb EW-19406-27 Mfr # PREPS-N-025
COMPARE
75 1/4" Hose Barb Polysulfone
$82.00USD / Each
PendoTech PRESS-N-050 Single-Use Pressure Sensor, Non-Sterile, Polycarbonate, 1/2" Hose Barb EW-19406-28 Mfr # PRESS-N-050
COMPARE
75 1/2" Hose Barb Polycarbonate
$84.00USD / Each
PendoTech PRESS-N-075 Single-Use Pressure Sensor, Non-Sterile, Polycarbonate, 3/4" Hose Barb EW-19406-29 Mfr # PRESS-N-075
COMPARE
75 3/4" Hose Barb Polycarbonate
$87.00USD / Each
PendoTech PRESS-N-100 Single-Use Pressure Sensor, Non-Sterile, Polycarbonate, 1" Hose Barb EW-19406-30 Mfr # PRESS-N-100
COMPARE
75 1" Hose Barb Polycarbonate
$75.00USD / Each
PendoTech PREPS-N-100 Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 1" Hose Barb EW-19406-31 Mfr # PREPS-N-100
COMPARE
75 1" Hose Barb Polysulfone
$121.00USD / Each
PendoTech PRESS-S-000 Single-Use Pressure Sensor, Sterile, Polycarbonate, Luer Fitting EW-19406-32 Mfr # PRESS-S-000
COMPARE
75 Female Luer Polycarbonate
$54.00USD / Each
PendoTech PREPS-N-012 Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 1/8" Hose Barb EW-19406-51 Mfr # PREPS-N-012
COMPARE
75 1/8" Hose Barb Polysulfone
$71.00USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 3/4" Sanitary Flange EW-19406-52 Mfr # PREPS-N-5-5
COMPARE
75 3/4" Sanitary Flange Inlet/Outlet Polysulfone
$100.00USD / Each
PendoTech PREPS-N-1-1 Single-Use Pressure Sensor, Polysulfone, Non-Sterile, 1" Sanitary Flange EW-19406-53 Mfr # PREPS-N-1-1
COMPARE
75 1" Sanitary Flange Inlet/Outlet Polysulfone
$120.00USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 1” Sanitary to 1” Hosebarb EW-19406-54 Mfr # PREPS-N-1-100
COMPARE
75 1” Sanitary Flange to 1” Hose Barb Polysulfone
$117.00USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 3/4" Sanitary Flange x 3/8” Hosebarb EW-19406-55 Mfr # PREPS-N-5-038
COMPARE
75 3/4" Sanitary Flange to 3/8” Hose Barb Polysulfone
$98.00USD / Each
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 3/4" Sanitary Flange x 1/2” Hosebarb EW-19406-56 Mfr # PREPS-N-5-050
COMPARE
75 3/4" Sanitary Flange to ½” Hose Barb Polysulfone
$98.00USD / Each
Showing 1- 18 of 18