Parker Hannifin Compression Fitting Ferrule Nuts
Get a secure seal at higher pressures
  • Constructed of durable stainless steel
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Compression nuts are required to create a proper seal.
$6.80 - $9.90USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Parker Hannifin Compression Fitting Ferrule Nut, 316 Stainless Steel, 1/16" OD
Buy More Save More
EW-03302-44 Mfr # 1 BZ-SS-C
COMPARE
1/16 Stainless Steel Compression Fitting Ferrule Nut, 316 Stainless Steel, 1/16" OD; 1/EA
$9.90USD / Each
Parker Hannifin Compression Fitting Ferrule Nut, 316 Stainless Steel, 1/8" OD
Buy More Save More
EW-03302-46 Mfr # 2 BZ-SS-C
COMPARE
1/8 Stainless Steel Compression Fitting Ferrule Nut, 316 Stainless Steel, 1/8" OD; 1/EA
$7.20USD / Each
Parker Hannifin Compression Fitting Ferrule Nut, 316 Stainless Steel, 1/4" OD
Buy More Save More
EW-03302-51 Mfr # 4 BZ-SS-C
COMPARE
1/4 Stainless Steel Compression Fitting Ferrule Nut, 316 Stainless Steel, 1/4" OD; 1/EA
$6.80USD / Each
Parker Hannifin Compression Fitting Ferrule Nut, 316 Stainless Steel, 3/8" OD
Buy More Save More
EW-03302-56 Mfr # 6 BZ-SS-C
COMPARE
3/8 Stainless Steel Compression Fitting Ferrule Nut, 316 Stainless Steel, 3/8" OD; 1/EA
$8.00USD / Each