Parker Balston Ultra High Purity Nitrogen Generator
Safely produce continuous supply of nitrogen gas
  • Compact design frees up laboratory space
  • Eliminates need for costly and dangerous high-pressure nitrogen cylinders
  • Offers long-term cost stability
Ultra-high purity (UHP) zero grade nitrogen generator transforms standard compressed air into 99.9999% nitrogen, exceeding the specification of UHP cylinder gas. The units can produce up to 1.1 lpm of UHP nitrogen gas and up to 2.0 lpm of research-grade purity nitrogen gas. A catalyst module is incorporated in the generator to oxidize hydrocarbons from the inlet air supply. Applications include GC carrier and make-up gas and low flow sample concentrators.
$18,196.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Power (Hz) Power (VAC)
Availability Pricing
Parker Hannifin Ultra High Purity Nitrogen Generator, 115 VAC EW-86442-15 Mfr # UHPN2-1100
COMPARE
1100 60 115
$18,196.00USD / Each