Lascar Electronics PanelPilotACE Touch-Screen Graphics Panel Meter
Create a personalized color interface without having to create lines of code
  • Meter connects to a PC via USB port for customization of color, units, alarms, input and output behaviors, and graphical or numerical representation of measurements
  • Free PanelPilotACE software download is compatible with Windows® XP, 2000, Vista and Windows 7
  • Four 16-bit bipolar analog inputs (scaleable using software to ±40V), eight digital I/O pins, and two alarm outputs (max current sink 10 mA)
  • RS-232, I2C, SPI connectivity, with expansion to include RS-485 and Ethernet already in the works
PanelPilotACE is a hardware and software platform providing you with the building blocks needed to create an industrial user interface. The Windows-based drag-and-drop software and library of predefined elements (such as meters, buttons, and switches) allow you to create your own content by combining elements and importing graphics, as well customizing color, text, units, and input scaling. Set up multiple layers of screens without needing multiple meters or having to write complex code. Software includes a "preview" function to emulate hardware inputs—letting you test setup before uploading to the meter.
$328.75 - $717.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type
Availability Pricing
Lascar Electronics SGD 43-A PanelPilotACE Graphics Meter, 4.3" TFT EW-18000-65 Mfr # SGD 43-A
COMPARE
4.3\" color touchscreen
$328.75USD / Each
Lascar Electronics SGD 70-A DK+ PanelPilotACE Development Kit, 7" TFT EW-18000-69 Mfr # SGD 70-A DK+
COMPARE
7\" color touchscreen
$717.00USD / Each