Ohaus SJX Scout Series Jewelry & Precious Metals Balances
Designed for a wide range of Legal-for-Trade applications—NTEP and Measurement Canada approved
  • Large backlit display lets you easily view results under any lighting conditions
  • Multiple application modes include parts counting, percent weighing, check weighing, totalization, and density determination
  • Weigh-below hook for density determination or specific gravity calculations
  • Quick stabilization time means fast results and increased efficiency
  • Superior overload protection safeguards balance from damage
  • Two hold modes—freeze the first stable reading or the first highest stable value
  • Stability indicator, auto tare, low-battery indicator, and auto shutoff features
  • Integrated security slot helps prevent theft
Designed for a wide range of Legal-for-Trade applications, the Class ll and Class III certified SJX Carat and Gold balances are ideal for weighing jewelry, precious metals and gemstones. Equipped with superior overload protection, large backlit display, fast stabilization time, multiple weighing units, application modes and connectivity options, these slim balances offer a truly user-friendly weighing experience.
ITEM(S) ON SALE
$720.09 - $1,048.14USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Description
Availability Pricing
Ohaus SJX323N/E Scout Carat 320ct x 0.01ct NTEP EW-11335-82 Mfr # SJX323N/E
COMPARE
64 Scout Carat 320ct x 0.01ct NTEP
$720.09USD / Each
$889.00USD / Each
Ohaus SJX622N/E Scout Gold 620g x 0.01g NTEP EW-11335-83 Mfr # SJX622N/E
COMPARE
620 Scout Gold 620g x 0.01g NTEP
$903.15USD / Each
$1,115.00USD / Each
Ohaus SJX1502N/E Scout Gold 1500g x 0.01g NTEP EW-11335-84 Mfr # SJX1502N/E
COMPARE
1500 Scout Gold 1500g x 0.01g NTEP
$1,048.14USD / Each
$1,294.00USD / Each
Ohaus SJX6201N/E Scout Gold 6200g x 0.1g NTEP EW-11335-85 Mfr # SJX6201N/E
COMPARE
6200 Scout Gold 6200g x 0.1g NTEP
$925.02USD / Each
$1,142.00USD / Each