This product is now discontinued, View Related Products
Ohaus SD Compact Bench / Shipping Scales
Designed for shipping, receiving, and general industrial weighing applications
  • Flexible mounting — remote indicator with coiled cable may be placed up to 9 feet away for eye-level mounting
  • Battery operable for greater portability — auto shutoff feature helps extend battery life
These bench scales feature a durable painted steel platform, remote indicator with two tactile function keys, battery operation, and flexible mounting capabilities. Ideal for home office, mail room, shipping and receiving, and general industrial weighing.
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus SD35 SD Shipping Scales, 35kg x 0.02kg (115 V) EW-11901-48 Mfr # SD35
COMPARE
35 77 115
$257.58USD / Each
$318.00USD / Each
Ohaus SD75 SD Shipping Scales, 75kg x 0.05kg (115 V) EW-11901-49 Mfr # SD75
COMPARE
75 165 115
Call For Price
Ohaus SD75L SD Shipping Scales, 75kg x 0.05kg, with Large Platform (115 V) EW-11901-50 Mfr # SD75L
COMPARE
75 165 115
Call For Price
Ohaus SD200 SD Shipping Scales, 200kg x 0.1kg (115 V) EW-11901-51 Mfr # SD200
COMPARE
200 440 115
Call For Price
Ohaus SD200L SD Shipping Scales, 200kg x 0.1kg, with Large Platform (115 V) EW-11901-52 Mfr # SD200L
COMPARE
200 440 115