Ohaus School and Primer Balances
The OHAUS SB1200 School Balance is the ideal teaching tool to introduce students to the science of metrology.
  • Interchangeable pans are removable for pouring, cleaning, and storing
  • Lightweight, yet durable plastic design
  • Dual pointers allow reading from both sides
  • Easy to use zero adjustment
  • So affordable, every student can have one
With a capacity of 2000 grams and a pivoting beam that provides accuracy to .5 grams, mass measurements can be achieved for small objects such as paper clips, marbles, and coins, as well as heavier objects such as books. This versatility makes this balance a valuable part of any science curriculum.
ITEM(S) ON SALE
$71.20USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus SB1200 School Balance, 2000g x 0.5G EW-11600-34 Mfr # SB1200
COMPARE
External 2000 0.5
$71.20USD / Each
$89.00USD / Each