Ohaus Explorer Toploading Balances
Ingenious modular design with improved performance
  • Modular desgin allows display to be separated from the weighing base
  • Optimized linearity and repeatability
  • Autocal™ automatic internal calibration ensures results are accurate
  • Intuitive operation with large, color touch screen display and icon-driven software
  • Easy to use SmarText™ graphical software
  • Metal base, ABS housing, and stainless steel pan
  • Color display and icon-driven navigation
  • Touchless sensors for hands-free operation
The OHAUS Explorer’s advanced applications simplify even the most complex laboratory measurements. Whether it’s determining the difference between initial and residual weights or calculating the density of solids and liquids, Explorer eliminates the need for time consuming manual calculations and data logging. The high resolution display and innovative user interface make balance setup and application use effortless.
ITEM(S) ON SALE
$2,610.96 - $5,095.50USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Ohaus EX223/E Explorer Toploading Balance, 220Gx0.001g, with External Calibration EW-11610-55 Mfr # EX223/E
COMPARE
220 100 to 240 0.001
$2,610.96USD / Each
$3,036.00USD / Each
Ohaus EX423/E Explorer Toploading Balance, 420Gx0.001g, with External Calibration EW-11610-56 Mfr # EX423/E
COMPARE
420 100 to 240 0.001
$3,156.20USD / Each
$3,670.00USD / Each
Ohaus EX2202/E Explorer Toploading Balance, 2200Gx0.01g, with External Calibration EW-11610-57 Mfr # EX2202/E
COMPARE
2200 100 to 240 0.01
$2,848.32USD / Each
$3,312.00USD / Each
Ohaus EX4202/E Explorer Toploading Balance, 4200Gx0.01g, with External Calibration EW-11610-58 Mfr # EX4202/E
COMPARE
4200 100 to 240 0.01
$3,320.46USD / Each
$3,861.00USD / Each
Ohaus EX6202/E Explorer Toploading Balance, 6200Gx0.01g, with External Calibration EW-11610-59 Mfr # EX6202/E
COMPARE
6200 100 to 240 0.01
$3,801.20USD / Each
$4,420.00USD / Each
Ohaus EX6201/E Explorer Toploading Balance, 6200Gx0.1g, with External Calibration EW-11610-61 Mfr # EX6201/E
COMPARE
6200 100 to 240 0.1
$3,052.14USD / Each
$3,549.00USD / Each
Ohaus EX423 Explorer Toploading Balance, 420g x 0.001g, with Internal Calibration EW-11800-04 Mfr # EX423
COMPARE
420 100 to 240 0.001
$3,557.82USD / Each
$4,137.00USD / Each
Ohaus EX623 Explorer Toploading Balance, 620g x 0.001g, with Internal Calibration EW-11800-05 Mfr # EX623
COMPARE
620 100 to 240 0.001
$4,347.30USD / Each
$5,055.00USD / Each
Ohaus EX1103 Explorer Toploading Balance, 1100g x 0.001g, with Internal Calibration EW-11800-06 Mfr # EX1103
COMPARE
1100 100 to 240 0.001
$5,095.50USD / Each
$5,925.00USD / Each
Ohaus EX2202 Explorer Toploading Balance, 2200g x 0.01g, with Internal Calibration EW-11800-07 Mfr # EX2202
COMPARE
2200 100 to 240 0.01
$3,290.36USD / Each
$3,826.00USD / Each
Ohaus EX4202 Explorer Toploading Balance, 4200g x 0.01g, with Internal Calibration EW-11800-08 Mfr # EX4202
COMPARE
4200 100 to 240 0.01
$3,782.28USD / Each
$4,398.00USD / Each
Ohaus EX6202 Explorer Toploading Balance, 6200g x 0.01g, with Internal Calibration EW-11800-09 Mfr # EX6202
COMPARE
6200 100 to 240 0.01
$4,292.26USD / Each
$4,991.00USD / Each
Ohaus EX10202 Explorer Toploading Balance, 10200g x 0.01g, with Internal Calibration EW-11800-10 Mfr # EX10202
COMPARE
10200 100 to 240 0.01
$5,091.20USD / Each
$5,920.00USD / Each
Ohaus EX6201 Explorer Toploading Balance, 6200g x 0.1g, with Internal Calibration EW-11800-11 Mfr # EX6201
COMPARE
6200 100 to 240 0.1
$3,569.00USD / Each
$4,150.00USD / Each
Ohaus EX10201 Explorer Toploading Balance, 10200g x 0.1g, with Internal Calibration EW-11800-12 Mfr # EX10201
COMPARE
10200 100 to 240 0.1
$3,910.42USD / Each
$4,547.00USD / Each
Showing 1- 15 of 15