Ohaus Explorer High Capacity Toploading Balances
The Explorer high-capacity balances are designed for heavy-duty applications where accuracy is a must.
  • Intelligent performance – optimized linearity and repeatability
  • Intelligent calibration - AutoCal automatic internal calibration ensures your results are accurate
  • Intuitive operation – Large color touch-screen display and icon-driven application software
  • Intuitive software - SmarText™ 2.0 is OHAUS’ easy-to-use graphical software
  • Practical touchless sensors - Hands-free operation
  • Ingenious modular design - Display can be separated from the weighing base for maximum flexibility in any work area
The OHAUS Explorer’s advanced applications simplify even the most complex laboratory measurements. Whether it’s determining the difference between initial and residual weights or calculating the density of solids and liquids, Explorer eliminates the need for time consuming manual calculations and data logging. The high resolution display and innovative user interface make balance setup and application use effortless.
ITEM(S) ON SALE
$4,322.36 - $6,183.40USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Ohaus EX12001 Explorer High Capacity Toploading/Bench Scale 12kg x 0.1g EW-11610-62 Mfr # EX12001
COMPARE
12000 100 to 240 0.1
$4,322.36USD / Each
$5,026.00USD / Each
Ohaus EX24001 Explorer High Capacity Toploading/Bench Scale 24kg x 0.1g EW-11610-63 Mfr # EX24001
COMPARE
24000 100 to 240 0.1
$5,264.92USD / Each
$6,122.00USD / Each
Ohaus EX35001 Explorer High Capacity Toploading/Bench Scale 35kg x 0.1g EW-11610-64 Mfr # EX35001
COMPARE
35000 100 to 240 0.1
$6,183.40USD / Each
$7,190.00USD / Each