Ohaus® Defender Multifunctional Scale Indicators
Designed to handle demanding industrial and commercial applications
  • Stores up to 50,000 items and 1,000 user profiles
  • View results clearly on large backlit LCD
  • NTEP Class III/IIIL certified
The multifunctional scale indicators are ideal for use in production, packaging, inventory, and shipping applications. Common functions include parts counting, checkweighing, percent weighing, and dynamic weighing. A large, backlit LCD quickly displays results in a choice of multiple weighing units (g, kg, lb, oz, lb:oz, metric tonne, and short ton). The alphanumeric keypad and function keys are simple to use, while on-screen prompts provide operational guidance.
Major features include maximum configurability print output, GMP/GLP data output, library/user management, and multiple connectivity options. Easily export weighing results through a standard RS-232 interface or a Micro SD slot. Other options include a second RS-232, RS-485 or USB device; Ethernet, and WiFi/Bluetooth® interface. Store up to 50,000 items and 1,000 user profiles on a MicroSD card (included).
Instantly connect the indicator to your bench or floor scale without any extra configuration by using EasyConnect™—saving you time and money.
ITEM(S) ON SALE
$717.66 - $882.90USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Power (VAC) Description
Availability Pricing
Ohaus TD52P Defender Multifunctional Scale Indicator for Standard Applications EW-11803-30 Mfr # TD52P
COMPARE
External, with calibration mass 100 to 240 Multifunctional Scale Indicator for Standard Applications, ABS Plastic
$717.66USD / Each
$886.00USD / Each
Ohaus TD52XW Defender Multifunctional Scale Indicator for Extreme Applications, IP68 EW-11803-31 Mfr # TD52XW
COMPARE
External, with calibration mass 100 to 240 Multifunctional Scale Indicator for Extreme Applications, Stainless Steel, IP68
$882.90USD / Each
$1,090.00USD / Each