This product is now discontinued, View Related Products
Oakton® Waterproof PC 650 pH/Conductivity Meters
Top-of-the-line dual parameter meters offer features typically found on much more expensive models!
  • Simultaneously measure your choice of pH, mV, or ISE and conductivity
  • Log up to 500 data points with GLP compliance
  • Wireless IrDA communication for truly waterproof interfacing capabilities
These 650 meters feature user-selectable “calibration due” alarms, automatic or manual standards recognition, and single or multiple calibration points — providing flexibility in meeting your quality objectives. The large backlit LCD shows the measured value, measured units, temperature, time, battery life, and menu options. Infrared (IrDA) wireless technology lets you easily download data without any cables.
PC 650 meters measure pH and ISE with a 0.001 maximum resolution. The PC 650 also measures conductivity, TDS, salinity, or resistivity using either a 2- or 4-electrode conductivity cell.
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Waterproof Meter with probes
Call For Price
COMPARE
Waterproof Meter with probes and NIST-traceable Calibration
Call For Price
COMPARE
Waterproof Handheld Meter Only
Call For Price
COMPARE
Waterproof Meter only with NIST-traceble Calibration
Call For Price
COMPARE
Waterproof Handheld Meter Kit
Call For Price
COMPARE
Waterproof Meter Kit with NIST-traceable Calibration