Oakton® Waterproof PC 450 pH/CON Portable Meters
Select meter with combination pH/conductivity/temperature probe or separate pH and conductivity/temperature probes
  • Intuitive, user-friendly icons—electrode status displays pH slope
  • Grip-Clip™ electrode holder quickly and easily secures electrode to side of beaker or container (for use with separate pH and conductivity/ATC probes)
  • Memory of 500 data sets and USB or RS-232 output for tranfer to PC
  • Extended battery life or optional universal power adapter
  • Built-in stand with wall mount allows for easy benchtop measurement or storage
9 PRODUCT OPTIONS
Compareitem specifications
Max Conductivity Max pH (pH) Description
Availability Pricing
COMPARE
200 mS 16 Waterproof Portable Meter with Combination Probe
$1,122.00USD / Each
$1,320.00USD / Each
COMPARE
200 mS 16 Waterproof Portable Meter with Combination Probe and Calibration
$1,464.00USD / Each
COMPARE
200 mS 16 Waterproof Portable Meter with Separate Probes
$1,164.50USD / Each
$1,370.00USD / Each
COMPARE
200 mS 16 Waterproof Portable Meter with Separate Probes and Calibration
$1,509.00USD / Each
COMPARE
200 mS 16 Waterproof Portable Meter Only
$930.75USD / Each
$1,095.00USD / Each
COMPARE
200 mS 16 Waterproof Kit with Combination Probe
$1,232.50USD / Each
$1,450.00USD / Each
COMPARE
200 mS 16 Waterproof Kit with Combination Probe and Calibration
$1,589.00USD / Each
COMPARE
200 mS 16 Waterproof Kit with Separate pH and Conductivity Probes
$1,283.50USD / Each
$1,510.00USD / Each
COMPARE
200 mS 16 WP Kit w/Separate pH and Conductivity Probes and Calibration
$1,631.00USD / Each
Showing 1- 9 of 9