This product is now discontinued, View Related Products
Oakton® Waterproof CD 650 Conductivity/Dissolved Oxygen Meters
Multiple parameter models for all of your conductivity/DO measurement needs
  • Programmable ''calibration due'' date for increased data integrity
  • Stores up to 500 data points with time-and-date stamp for GLP compliance
  • Wirelessly download data via IrDA port
These top-of-the-line waterproof meters offer single or multiparameter measurement capabilities to meet all of your testing needs. The IP67-rated housing allows use in dirty and wet environments without harm to the meter. Store up to 500 data points and download to computer or printer with the infrared (IrDA) wireless technology — no need for additional cords. User-selectable “calibration due” and set point alarms, automatic or manual standards recognition, and single- or multiple-point calibration provide flexibility in meeting you data quality objectives. The large backlit, graphic LCD shows the measured value and units, temperature, time, battery life, and menu options.
6 PRODUCT OPTIONS
Compareitem specifications
Accessories Included NIST Calibration Included Probe Included
Availability Pricing
COMPARE
No No Yes
Call For Price
COMPARE
No Yes Yes
Call For Price
COMPARE
No No No
Call For Price
COMPARE
No Yes No
Call For Price
COMPARE
Yes No Yes
Call For Price
COMPARE
Yes Yes Yes