Oakton pH Indicator Strips
Vivid and easy-to-distinguish colors at every pH interval
Convenient flip-top vials include color comparator chart. Read results immediately after exposing test area to the solution. Strips measure 2-3/4"L x 1/4"W (7 x 2 cm). Each vial contains 100 strips.
$30.50USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Measurement Points
Availability Pricing
COMPARE
0 to 14 pH in 1 pH unit increments
$30.50USD / Pkg of 100
COMPARE
5 to 9 pH in 0.5 pH unit increments
$30.50USD / Pkg of 100