Oakton® pH Electrode Storage and Cleaning Solutions
Keep your pH probes clean and properly stored for longevity
  • Easily remove grease, protein, and other deposits with cleaning solution
  • Make electrode last longer— store in our storage solution
  • Choose 500-mL bottle or case of 24 500-mL bottles
Remove grease and oil deposits from your electrodes with our cleaning solution. Cleaning solution contains pepsin. After cleaning, keep your electrodes in proper working condition with our storage solution.
$18.75 - $478.00USD / Pint of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Volume (oz) Description
Availability Pricing
COMPARE
500 16 Electrode Storage Solution; 500 mL
$24.50USD / Pint of 1
COMPARE
500 16 Electrode Cleaning Solution; 500 mL
$18.75USD / Pint of 1
COMPARE
500 16.9 Electrode Storage Solution; 24 x 500 mL Bottles/Cs
$478.00USD / Case of 24