Oakton® pH Buffer Pouches
Economical pouches are accurate and convenient
  • Single-use pouch—just tear, dip, calibrate
  • Standardized to NIST-traceable pH references
  • Sold in pack of 20 pouches
Buffers are traceable and offer ±0.01 accuracy at 77°F (25°C). Pack of 20 pouches.
$39.75 - $47.00USD / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Buffer Kit Values Description
Availability Pricing
COMPARE
4.01 pH Calibration Buffer Pouches, pH 4.01, 20/Pk
$39.75USD / Pkg of 20
COMPARE
7.00 pH Calibration Buffer Pouches, pH 7.00, 20/Pk
$42.75USD / Pkg of 20
COMPARE
10.00 pH Calibration Buffer Pouches, pH 10.00, 20/Pk
$42.75USD / Pkg of 20
COMPARE
4.01, 7.00, and 10.00 pH Calibration Buffer Pouches, Assorted; 20/Pk
$47.00USD / Pkg of 20