Oakton® pH 6+ Handheld pH Meters
High +/-0.01 pH accuracy at an economical price
  • Convenient, compact design
  • Up to five-point push-button calibration with autobuffer recognition
  • Displays pH or mV and temperature with the press of a button
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC)ensures accuracy from 0 to 100°C. Meters include a protective rubber boot that also acts as a meter stand.
ITEM(S) ON SALE
$263.75 - $605.25USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max pH (pH) Min pH (pH) Description
Availability Pricing
COMPARE
14 0 Handheld Meter with ATC Probe; Order pH probe separately
$263.75USD / Each
$310.30USD / Each
COMPARE
14 0 Handheld Meter with pH Probe
$316.54USD / Each
$372.40USD / Each
COMPARE
14 0 Handheld Meter Kit with Case, Solutions, and pH/ATC Probe
$388.62USD / Each
$457.20USD / Each
COMPARE
14 0 Handheld Meter with NIST-traceable Calibration
$458.30USD / Each
COMPARE
14 0 Handheld Meter with pH Probe and NIST-traceable Calibration
$520.50USD / Each
COMPARE
14 0 Handheld Meter Kit with Case, Solutions, pH/ATC Probe, and NIST-traceable Calibration
$605.25USD / Each