Oakton® PH 550 Benchtop pH Meter Kits
Economical meter for benchtop pH measurement
  • Automatically recognizes calibration solutions
  • Compact design saves valuable benchtop space
  • Data logging allows you to store and recall up to 50 groups of data
  • IP54-rated housing is dust- and spill-proof
The PH 550 benchtop pH meter features a microprocessor chip which provides automatic temperature compensation, data storage, parameter setup, and Max/Min reading display. The meter offers a pH range from 0 to 14 and a three-point automatic calibration, with the ability to recognize standard buffer solutions (USA or NIST series). The advanced digital processing technology improves meter response time and measuring accuracy. A stable reading display icon appears when the measuring value is stable.
The 3-in-1 combination pH electrode measures pH and temperature simultaneously. The compact meter includes a detachable electrode holder and three bottles of pH buffer solution. It has a dustproof and spillproof rating of IP54. Connectors are protected by silicone seal caps.
$529.00 - $609.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Benchtop pH Meter Kit with Probe, Stand, and pH Buffers
$609.00USD / Pkg of 1
COMPARE
Benchtop pH Meter Kit with Probe and Stand
$529.00USD / Each