Oakton® pH 5+ Handheld pH Meters
High +/-0.01 pH accuracy at an economical price
  • Convenient, compact design
  • Up to five-point push-button calibration with autobuffer recognition
  • Displays pH and temperature with the press of a button
These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC) ensures accuracy from 0 to 80°C. Meters include a protective rubber boot that also acts as a meter stand.
ITEM(S) ON SALE
$340.00 - $635.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Handheld Meter with ATC; Order pH probe separately
$340.00USD / Each
$400.00USD / Each
COMPARE
meter only with NIST-traceable Calibration
$474.00USD / Each
COMPARE
Handheld Meter with pH Probe
$430.95USD / Each
$507.00USD / Each
COMPARE
meter with probe with NIST-traceable Calibration
$529.00USD / Each
COMPARE
Handheld Meter Kit w/ Case, Solutions, pH/ATC Probe
$510.00USD / Each
$600.00USD / Each
COMPARE
Meter kit with NIST-traceable Calibration
$635.00USD / Each