Oakton® High-Accuracy pH Buffer Solutions

pH Calibration buffers for tight tolerances
  • For meters with 0.001 resolution
Oakton® High-accuracy pH buffer solutions are sold in 500 mL glass bottles, individually or as a case of twelve.
Buffers are color-coded for easy visual identification.
$43.00 - $44.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Buffer Value Color Volume (mL)
Availability Pricing
COMPARE
4 Red 500
$43.00USD / Each
COMPARE
7 Yellow 500
$43.00USD / Each
COMPARE
10 Blue 500
$44.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat